Learn More
Apexbio Technology LLC HyperScribe T7 High Yield RNA Synthesis Kit Plus 100 rxns
SDP
Catalog No. 503107141
Encompass_Preferred
HyperScribeTM T7 High Yield RNA Synthesis Kit Plus utilizes T7 RNA polymerase to conduct rapid in vitro RNA transcription reactions This kit is designed for the generation of high-yield RNA transcripts highly specific radioactively labeled RNA probes or RNA incorporating modified nucleotides such as capped dye-labeled or biotinylated RNA The kit is suitable for numerous applications including in vitro translation antisense RNA and RNAi experiments RNA vaccines investigation of RNA structure and function ribozyme biochemistry RNase protein assays and probe-based hybridization blotting Each standard reaction volume is 20 L and kits are provided with sufficient reagents for 25/50/100 reactions Using 1 g of the provided control template per standard reaction yields of up to 180 g RNA per reaction can be achieved Accordingly the 25/50/100-reaction kits can produce up to 4 5 mg/9 mg/18 mg RNA respectively
Catalog No. 50-310-7141 Supplier Apexbio Technology LLC Supplier No. K1401100
HyperScribeTM T7 High Yield RNA Synthesis Kit Plus utilizes T7 RNA polymerase to conduct rapid in vitro RNA transcription reactions This kit is designed for the generation of high-yield RNA transcripts highly specific radioactively labeled RNA probes or RNA incorporating modified nucleotides such as capped dye-labeled or biotinylated RNA The kit is suitable for numerous applications including in vitro translation antisense RNA and RNAi experiments RNA vaccines investigation of RNA structure and function ribozyme biochemistry RNase protein assays and probe-based hybridization blotting Each standard reaction volume is 20 L and kits are provided with sufficient reagents for 25/50/100 reactions Using 1 g of the provided control template per standard reaction yields of up to 180 g RNA per reaction can be achieved Accordingly the 25/50/100-reaction kits can produce up to 4 5 mg/9 mg/18 mg RNA respectively