Learn More
LGC GENOMICS LLC (Lucigen) T7 R&DNA Polymerase, 5,000 U @ 50 U/LIncorporate both dNTPs and standard NTPs into RNA transcripts with this unique, mutated T7 RNA polymerase.
Catalog No. NC1579358
Encompass
T7 R&DNA™ Polymerase is a recombinant mutant form of T7 RNA polymerase (Y639F mutant), in which Tyr-639 has been replaced by Phe.
Catalog No. NC1579358 Supplier LGC GENOMICS LLC Supplier No. D7P9205K
T7 R&DNA™ Polymerase is a recombinant mutant form of T7 RNA polymerase (Y639F mutant), in which Tyr-639 has been replaced by Phe.