Learn More
New England Biolabs, Inc. T7 RNA Polymerase – 25000 units
SDP
Catalog No. 50811666
Encompass
Bacteriophage T7 RNA Polymerase is a DNA-dependent RNA polymerase that is highly specific for the T7 phage promoters. The 99 KD enzyme catalyzes in vitro RNA synthesis from a cloned DNA sequence under the T7 promoters. RNA produced using the T7 RNA Polymerase is suitable for many applications in research and biotechnology. Reaction Conditions 1X RNAPol Reaction Buffer, supplemented with 0.5 mM each ATP, UTP, GTP, CTP, and DNA template containing the T7 RNA Polymerase promoter. Incubate at 37°C.
Catalog No. 50-811-666 Supplier New England Biolabs, Inc. Supplier No. M0251L
Bacteriophage T7 RNA Polymerase is a DNA-dependent RNA polymerase that is highly specific for the T7 phage promoters. The 99 KD enzyme catalyzes in vitro RNA synthesis from a cloned DNA sequence under the T7 promoters. RNA produced using the T7 RNA Polymerase is suitable for many applications in research and biotechnology. Reaction Conditions 1X RNAPol Reaction Buffer, supplemented with 0.5 mM each ATP, UTP, GTP, CTP, and DNA template containing the T7 RNA Polymerase promoter. Incubate at 37°C.