Learn More
Sino Biological T7 RNA Polymerase 25 kU
Catalog No. 5000696179
Encompass_Preferred
T7 RNA Polymerase is a DNA-dependent 5‘→3’ RNA polymerase that exhibits high specificity for the T7 promoter sequence(5‘-TAATACGACTCACTATAGGGAGA-3’). It catalyzes the incorporation of nucleoside triphosphates (NTPs) downstream of a T7 promoter on either single-stranded or double-stranded DNA templates, synthesizing RNA complementary to the template strand.T7 RNA Polymerase is capable of utilizing modified NTPs, such as those labeled with biotin, digoxigenin, or fluorescein, enabling the synthesis of various labeled RNA molecules. It also demonstrates high specificity for the T7 promoter.
Catalog No. 50-006-96179 Supplier Sino Biological Supplier No. 962T7RP003025
T7 RNA Polymerase is a DNA-dependent 5‘→3’ RNA polymerase that exhibits high specificity for the T7 promoter sequence(5‘-TAATACGACTCACTATAGGGAGA-3’). It catalyzes the incorporation of nucleoside triphosphates (NTPs) downstream of a T7 promoter on either single-stranded or double-stranded DNA templates, synthesizing RNA complementary to the template strand.T7 RNA Polymerase is capable of utilizing modified NTPs, such as those labeled with biotin, digoxigenin, or fluorescein, enabling the synthesis of various labeled RNA molecules. It also demonstrates high specificity for the T7 promoter.