Learn More
INTACT GENOMICS INC T7 RNA Polymerase 25,000 Units
Catalog No. 5000083067
Encompass
T7 RNA Polymerase is useful for sgRNA and mRNA synthesis, RNA synthesis from DNA templates and applications in mRNA therapeutics.Intact Genomics IG T7 RNAP is a purified recombinant T7 bacteriophage enzyme that is expressed and purified from E. coli. This enzyme is designed to perform in vitro transcription of a gene under a phage T7 promoter. The physical purity of this enzyme is ≥85% as assessed by SDS-PAGE with Coomassie blue staining and gel densitometry.Applications: Radiolabeled RNA probe preparation RNA synthesis from DNA template sgRNA and mRNA synthesis Applications in mRNA therapeutics
Catalog No. 50-000-83067 Supplier INTACT GENOMICS INC Supplier No. 3295
T7 RNA Polymerase is useful for sgRNA and mRNA synthesis, RNA synthesis from DNA templates and applications in mRNA therapeutics.Intact Genomics IG T7 RNAP is a purified recombinant T7 bacteriophage enzyme that is expressed and purified from E. coli. This enzyme is designed to perform in vitro transcription of a gene under a phage T7 promoter. The physical purity of this enzyme is ≥85% as assessed by SDS-PAGE with Coomassie blue staining and gel densitometry.Applications: Radiolabeled RNA probe preparation RNA synthesis from DNA template sgRNA and mRNA synthesis Applications in mRNA therapeutics