Learn More
Aldevron T7 RNA Polymerase, 2000ug
Catalog No. 503073862
Encompass_Preferred
2000ug dispense of wild type T7 RNA Polymerase. T7 RNA Polymerase catalyzes synthesis of RNA in the presence of a DNA template containing the T7 phage promoter.
Catalog No. 50-307-3862 Supplier Aldevron Supplier No. 91352000MCG
2000ug dispense of wild type T7 RNA Polymerase. T7 RNA Polymerase catalyzes synthesis of RNA in the presence of a DNA template containing the T7 phage promoter.