Learn More
Aldevron T7 RNA Polymerase, 130ug
Catalog No. 503073888
Encompass_Preferred
130ug dispense of wild type T7 RNA Polymerase. T7 RNA Polymerase catalyzes synthesis of RNA in the presence of a DNA template containing the T7 phage promoter.
Catalog No. 50-307-3888 Supplier Aldevron Supplier No. 9135130MCG
130ug dispense of wild type T7 RNA Polymerase. T7 RNA Polymerase catalyzes synthesis of RNA in the presence of a DNA template containing the T7 phage promoter.