Learn More
Sino Biological Anti-T7 RNA polymerase Antibody
Supplier:  Sino Biological 68171T4820
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant T7 RNA Polymerase. The specific IgG was purified by T7 RNA Polymerase affinity chromatography.
Catalog No. 50-237-3121