Learn More
Apexbio Technology LLC HyperScribe T7 High Yield RNA Synthesis Kit (N1-Methylpseudo-UTP) 25 rxns
SDP
Catalog No. 503107136
Encompass_Preferred
Our product the T7 High Yield RNA Synthesis Kit is based on the principle of T7 RNA polymerase and enables efficient in vitro RNA transcription in a relatively short period of time The kit includes the modified nucleotide N1-Methylpseudo-UTP (m1 ) (Catalog B8049) which can replace regular UTP to reduce host cell immune responses and improve RNA stability This kit is designed to produce high-yield RNA transcripts highly specific radiolabeled RNA probes as well as capped RNA dye-labeled or biotin-labeled modified nucleotide RNAs RNAs synthesized by this kit have broad applications in biological experiments such as in vitro translation antisense RNA and RNAi experiments RNA vaccine development RNA structural and functional studies ribozyme biochemistry RNase protein assays and probe-based hybridization blots This kit supports for 25/50/100 reactions at 20 L With 1 g of control template can yield up to 180 g of RNA
Catalog No. 50-310-7136 Supplier Apexbio Technology LLC Supplier No. K1417100
Our product the T7 High Yield RNA Synthesis Kit is based on the principle of T7 RNA polymerase and enables efficient in vitro RNA transcription in a relatively short period of time The kit includes the modified nucleotide N1-Methylpseudo-UTP (m1 ) (Catalog B8049) which can replace regular UTP to reduce host cell immune responses and improve RNA stability This kit is designed to produce high-yield RNA transcripts highly specific radiolabeled RNA probes as well as capped RNA dye-labeled or biotin-labeled modified nucleotide RNAs RNAs synthesized by this kit have broad applications in biological experiments such as in vitro translation antisense RNA and RNAi experiments RNA vaccine development RNA structural and functional studies ribozyme biochemistry RNase protein assays and probe-based hybridization blots This kit supports for 25/50/100 reactions at 20 L With 1 g of control template can yield up to 180 g of RNA