Learn More
Creative Diagnostics T7 RNA Polymerase ELISA Kit, 96T, Sandwich-ELISA, Detection Limit <0.1 ng/mL, Lower Limit of Quantitation 0.25 ng/mL, Intra-assay CV ≤10%, Inter-assay CV ≤15%, 450 nm, For research use only
Supplier:  Creative Diagnostics DEIANS032
Encompass
T7 RNA Polymerase ELISA Kit, 96T, Sandwich-ELISA, Detection Limit <0.1 ng/mL, Lower Limit of Quantitation 0.25 ng/mL, Intra-assay CV ≤10%, Inter-assay CV ≤15%, 450 nm, For research use only
Catalog No. NC2468389