Learn More
Profoldin T7 RNA Polymerase Assay Kit, 10x Buffer 400 μL, 100x DNA 33 μL, 100x NTPs 33 μL, 2x Dye 1550 μL, 50 mM EDTA 1550 μL, 384-well plate, 96-well plate, 100 assays
Catalog No. NC2148368
Encompass
T7 RNA Polymerase Assay Kit, 10x Buffer 400 μL, 100x DNA 33 μL, 100x NTPs 33 μL, 2x Dye 1550 μL, 50 mM EDTA 1550 μL, 384-well plate, 96-well plate, 100 assays
This item is not returnable. View return policy
Catalog No. NC2148368 Supplier Profoldin Supplier No. T7RPA100K
T7 RNA Polymerase Assay Kit, 10x Buffer 400 μL, 100x DNA 33 μL, 100x NTPs 33 μL, 2x Dye 1550 μL, 50 mM EDTA 1550 μL, 384-well plate, 96-well plate, 100 assays