Learn More
Tecan D300e T8+ Cassettes
Supplier:  Tecan 30097370
Encompass
For use with D300e Digital Dispenser. For non contact dispensing of DMSO and aqueous solutions with a surfactant from 11 pL to 10 µL. 20 cassettes/sales unit. With the Tecan® D300e Digital Dispenser, you can automatically dispense any volume into any well of any assay plate, quickly and easily. Simply add your liquid tothe Dispensehead and let the instrument do the rest.
Catalog No. NC0846635