Learn More
Tecan T8+ Dispensehead Cassettes, 20 per box
Catalog No. NC3823917
Encompass_Preferred
• D300e Dispensehead Cassettes; DUO Dispensehead Cassettes
• 8 Dispenseheads
• 11 pl to 10 µL
• 20 Cassettes Per Box
T8+ Dispensehead Cassettes are designed for the Duo Digital Dispenser, enabling precise non-contact dispensing of DMSO or aqueous solutions with surfactant in volumes ranging from 11 pl to 10 µl. Effortlessly dispense directly into 12- to 1536-well plates. Simply add your liquid (up to 20 µl) to the T8+ Dispensehead and let the instrument handle the rest. These cassettes are compatible with both the Duo Digital Dispenser and the D300e Digital Dispenser.
Catalog No. NC3823917 Supplier Tecan Supplier No. 30253252
• D300e Dispensehead Cassettes; DUO Dispensehead Cassettes
• 8 Dispenseheads
• 11 pl to 10 µL
• 20 Cassettes Per Box
T8+ Dispensehead Cassettes are designed for the Duo Digital Dispenser, enabling precise non-contact dispensing of DMSO or aqueous solutions with surfactant in volumes ranging from 11 pl to 10 µl. Effortlessly dispense directly into 12- to 1536-well plates. Simply add your liquid (up to 20 µl) to the T8+ Dispensehead and let the instrument handle the rest. These cassettes are compatible with both the Duo Digital Dispenser and the D300e Digital Dispenser.