Learn More
ATCC T84; Colon Carcinoma; Human (Homo sapiens)
Catalog No. 502382858
Encompass
T84 is a transplantable human carcinoma cell line isolated from the lung metastasis of a 72-year-old, male, colon cancer patient.
Catalog No. 50-238-2858 Supplier ATCC Supplier No. CCL248
T84 is a transplantable human carcinoma cell line isolated from the lung metastasis of a 72-year-old, male, colon cancer patient.