Learn More
Medchemexpress LLC HY-32219 5mg Medchemexpress, T863 CAS:701232-20-4 Purity:>98%
SDP
Catalog No. 502029307
Encompass_Preferred
Medchemexpress, HY-32219 5mg T863 CAS:701232-20-4 T863 is an orally active, selective and potent DGAT1 (Acyl-CoA:diacylglycerol acyltransferase 1) inhibitor that interacts with the acyl-CoA binding site of DGAT1, and inhibits triacylglycerol synthesis in cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9307 Supplier Medchemexpress LLC Supplier No. HY322195MG
Medchemexpress, HY-32219 5mg T863 CAS:701232-20-4 T863 is an orally active, selective and potent DGAT1 (Acyl-CoA:diacylglycerol acyltransferase 1) inhibitor that interacts with the acyl-CoA binding site of DGAT1, and inhibits triacylglycerol synthesis in cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.