Learn More
ATCC T98G; Glioblastoma Multiforme; Human (Homo sapiens)
Catalog No. 502383538
Encompass
T98G [T98-G] is a fibroblast-like cell that was isolated from the brain of a glioblastoma multiforme White, 61-year-old, male patient. This cell line was deposited by GH Stein and can be used in neuroscience research.
Catalog No. 50-238-3538 Supplier ATCC Supplier No. CRL1690
T98G [T98-G] is a fibroblast-like cell that was isolated from the brain of a glioblastoma multiforme White, 61-year-old, male patient. This cell line was deposited by GH Stein and can be used in neuroscience research.