Learn More
Medchemexpress LLC HY-100114 50mg Medchemexpress, TA-01 CAS:1784751-18-3 Purity:>98%
SDP
Catalog No. 501931719
Encompass_Preferred
Medchemexpress, HY-100114 50mg TA-01 CAS:1784751-18-3 TA-01 is a potent CK1 and p38 MAPK inhibitor, with IC 50 s of 6.4 nM, 6.8 nM, 6.7 nM for CK1ε, CK1δ and p38 MAPK, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1719 Supplier Medchemexpress LLC Supplier No. HY10011450MG
Medchemexpress, HY-100114 50mg TA-01 CAS:1784751-18-3 TA-01 is a potent CK1 and p38 MAPK inhibitor, with IC 50 s of 6.4 nM, 6.8 nM, 6.7 nM for CK1ε, CK1δ and p38 MAPK, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.