Learn More
Medchemexpress LLC TA-02 | 1784751-19-4 | 99.9% | 333.33 g/mol | C20H13F2N3 | 1 ML
SDP
Catalog No. 5000215458
Encompass_Preferred
TA-02 is a small-molecule inhibitor of p38 MAPK and an analog of SB 203580, reported to show potent biochemical activity (IC50 ≈ 20 nM). The compound is supplied with high purity and is offered in ready-to-use solution formats as well as solid quantities for research use in biochemical and cell-based assays.
  • Potent p38 MAPK inhibition (IC50 ≈ 20 nM).
  • High purity, suitable for research applications.
  • Available as a 10 mM solution in DMSO for immediate use.
  • Solid form available for custom dilution and formulation.
  • Stable under recommended storage conditions for reliable results.
Catalog No. 50-002-15458 Supplier Medchemexpress LLC Supplier No. HY10011510MM 1ML
TA-02 is a small-molecule inhibitor of p38 MAPK and an analog of SB 203580, reported to show potent biochemical activity (IC50 ≈ 20 nM). The compound is supplied with high purity and is offered in ready-to-use solution formats as well as solid quantities for research use in biochemical and cell-based assays.
  • Potent p38 MAPK inhibition (IC50 ≈ 20 nM).
  • High purity, suitable for research applications.
  • Available as a 10 mM solution in DMSO for immediate use.
  • Solid form available for custom dilution and formulation.
  • Stable under recommended storage conditions for reliable results.