Learn More
Medchemexpress LLC TA-02 | 1784751-19-4 | 99.9% | 333.33 g/mol | C20H13F2N3 | 10 MG
SDP
Catalog No. 5000215179
Encompass_Preferred
This small-molecule research compound is a potent p38 MAPK inhibitor reported to have an IC50 of 20 nM and additional activity against TGFBR-2. It is supplied as a high-purity solid or DMSO solution for biochemical and cellular studies.
  • Potent p38 MAPK inhibition (IC50 = 20 nM).
  • Also inhibits TGFBR-2.
  • High purity (99.9%).
  • Available as small solid quantities (e.g., 10 mg) and as a DMSO solution.
  • Suitable for biochemical and cellular assay applications.
Catalog No. 50-002-15179 Supplier Medchemexpress LLC Supplier No. HY10011510MG
This small-molecule research compound is a potent p38 MAPK inhibitor reported to have an IC50 of 20 nM and additional activity against TGFBR-2. It is supplied as a high-purity solid or DMSO solution for biochemical and cellular studies.
  • Potent p38 MAPK inhibition (IC50 = 20 nM).
  • Also inhibits TGFBR-2.
  • High purity (99.9%).
  • Available as small solid quantities (e.g., 10 mg) and as a DMSO solution.
  • Suitable for biochemical and cellular assay applications.