Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ta-02 | 1784751-19-4 | 99.9% | 333.33 g/mol | C20H13F2N3 | 100 MG
SDP

Catalog No. 5000215508
Encompass_Preferred

TA-02 is a small-molecule p38 MAPK inhibitor for research use. It is reported to inhibit p38 MAPK with an IC50 of about 20 nM and also shows inhibitory activity against TGFBR-2. The compound is supplied as a solid and as a DMSO solution for in vitro experimental applications.

  • Purity 99.85%.
  • Molecular weight 333.33 g/mol.
  • Molecular formula C20H13F2N3.
  • CAS number 1784751-19-4.
  • Available as solid (multiple sizes) and 10 mM solution in DMSO.
  • Intended for research and analytical applications (not for human use).

Catalog No. 50-002-15508 Supplier Medchemexpress LLC Supplier No. HY100115100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TA-02 is a small-molecule p38 MAPK inhibitor for research use. It is reported to inhibit p38 MAPK with an IC50 of about 20 nM and also shows inhibitory activity against TGFBR-2. The compound is supplied as a solid and as a DMSO solution for in vitro experimental applications.

  • Purity 99.85%.
  • Molecular weight 333.33 g/mol.
  • Molecular formula C20H13F2N3.
  • CAS number 1784751-19-4.
  • Available as solid (multiple sizes) and 10 mM solution in DMSO.
  • Intended for research and analytical applications (not for human use).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.