Learn More
Selleck Chemical LLC TA-02
SDP
Catalog No. 5000002328
Encompass_Preferred
TA-02 is a p38 MAPK inhibitor with IC50 of 20 nM TA-02 especially inhibits TGFBR-2
Catalog No. 50-000-02328 Supplier Selleck Chemical LLC Supplier No. S680725MG
TA-02 is a p38 MAPK inhibitor with IC50 of 20 nM TA-02 especially inhibits TGFBR-2