Learn More
GA International Cryogenic Tape for Laboratory Use 0.5" x 50' / 12.7mm x 15m Yellow 1" / 25.4mm From -196°C to +110°C (-321°F to +230°F)
Catalog No. NC1751889
Encompass_Preferred
Continuous writable tape for cryogenic use. Water-proof, resistant to liquid nitrogen, dry ice, multiple freeze-thaw cycles, and other harsh laboratory environments. Extra Strong CryoMax™ adhesive. Available in a variety of colors for freezer and liquid nitrogen use (as low as -196°C) on plastic and glass containers.
Catalog No. NC1751889 Supplier GA International Supplier No. TJTA13C150YE
Continuous writable tape for cryogenic use. Water-proof, resistant to liquid nitrogen, dry ice, multiple freeze-thaw cycles, and other harsh laboratory environments. Extra Strong CryoMax™ adhesive. Available in a variety of colors for freezer and liquid nitrogen use (as low as -196°C) on plastic and glass containers.