Learn More
TA Instruments TA Instruments, Discovery SA, Autosampler Tray, 10 Position, for TGA Hemispherical Quartz Sample Pans, Included with Discovery SA
Catalog No. 5000580628 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Discovery SA Autosampler Tray holds 10 positions for TGA hemispherical quartz sample pans. It is included with the Discovery SA and Q5000 SA and is available as an option for the Q5000 IR.
Catalog No. 50-005-80628 Supplier TA Instruments Supplier No. 957216.901
Waters TA Instruments Discovery SA Autosampler Tray holds 10 positions for TGA hemispherical quartz sample pans. It is included with the Discovery SA and Q5000 SA and is available as an option for the Q5000 IR.