Learn More
TA Instruments TA Instruments, HA Aluminum, Double Gap Rotor, Inner Radius 20.38 mm, Outer Radius 21.96 mm, Height 59.5 mm, HR X0, Smart Swap 2
Catalog No. 5000580763 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, Smart Swap 2, HA Aluminum, Double Gap Rotor has an inner radius of 20.38 mm, outer radius of 21.96 mm, and height of 59.5 mm.
Catalog No. 50-005-80763 Supplier TA Instruments Supplier No. 546021.941
Waters TA Instruments HR X0, Smart Swap 2, HA Aluminum, Double Gap Rotor has an inner radius of 20.38 mm, outer radius of 21.96 mm, and height of 59.5 mm.