Learn More
TA Instruments TA Instruments, Loop Decoupler Kit, for Use with Discovery TGA, Discovery SA, Connects Thermobalance Arms to Sample and Reference Hangdown Wires
Catalog No. 5000580742 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments Loop Decoupler Kit is designed for all Discovery TGA and Discovery SA instruments. It connects the thermobalance arms to sample and reference hangdown wires to ensure accurate measurements.
Catalog No. 50-005-80742 Supplier TA Instruments Supplier No. 957515.901
Waters TA Instruments Loop Decoupler Kit is designed for all Discovery TGA and Discovery SA instruments. It connects the thermobalance arms to sample and reference hangdown wires to ensure accurate measurements.