Learn More
TA Instruments TA Instruments, Tray Autosampler, TGA, for Discovery SA, TGA 550, TGA 5500, Discovery TGA 1st Gen, for TGA Platinum, Ceramic, Aluminum Punched Pan Sample Pans
Catalog No. 5000580631 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Tray Autosampler TGA is designed for use with Discovery SA, Q5000, TGA 550/5500, Discovery TGA 1st Gen., and Q5000 IR instruments. It accommodates TGA platinum, ceramic, and aluminum punched pan sample pans for automated analysis.
Catalog No. 50-005-80631 Supplier TA Instruments Supplier No. 957099.901
Waters TA Instruments Tray Autosampler TGA is designed for use with Discovery SA, Q5000, TGA 550/5500, Discovery TGA 1st Gen., and Q5000 IR instruments. It accommodates TGA platinum, ceramic, and aluminum punched pan sample pans for automated analysis.