Learn More
Strem, An Ascensus Company CAS# 69039-11-8. 1g. t-Butylimidotris(dimethylamino)tantalum(V), min. 98%. MFCD05863998
SDP
Supplier:  Strem, An Ascensus Company 7307001G
Encompass
CAS# 69039-11-8. 1g. t-Butylimidotris(dimethylamino)tantalum(V), min. 98%. MFCD05863998. Molecular Weight: 384.30. Molecular Formula: C10H27N4Ta. Color/form: pale-yellow solid Strem# 73-0700. http://www.strem.com/catalog/v/73-0700/
Catalog No. 50-204-5375