Learn More
MOLTOX Molecular Toxicology Inc STDisc TA102, 10/vial
SDP
Catalog No. NC1762612
Encompass
Consists of lyophilized & stablilized S.Typhimurium strain TA1535 cells. For use as culture inoculum.
Catalog No. NC1762612 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 71102.5L
Consists of lyophilized & stablilized S.Typhimurium strain TA1535 cells. For use as culture inoculum.