Learn More
Aviva Systems Biology Corp TAAR6 Antibody - C-terminal region (ARP65391_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65391_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TAAR6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human TAAR6. Alternate names: RP11-295F4.3, TA4, TAR4, TAR6, TRAR4ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-492