Learn More
Medchemexpress LLC Tabersonine | 4429-63-4 | MFCD28140545 | 98.0% | 10 MG
SDP
Catalog No. 5000295823
Encompass_Preferred
Tabersonine is a monoterpenoid indole alkaloid used as a research reagent. It has been reported as an NLRP3 inhibitor and is used in studies of inflammation, apoptosis, and neurodegeneration. Supplied as a 10 MG solid, it is intended for laboratory research use only and not for human or clinical applications.
  • Monoterpenoid indole alkaloid with reported NLRP3 inhibitory activity.
  • Useful for studying inflammation, apoptosis, and neurodegenerative processes.
  • Molecular formula C21H24N2O2; molecular weight 336.43 g·mol-1.
  • Supplied as a 10 MG solid for laboratory research.
  • For research use only, not for human or clinical use.
Catalog No. 50-002-95823 Supplier Medchemexpress LLC Supplier No. HYN143110MG
Tabersonine is a monoterpenoid indole alkaloid used as a research reagent. It has been reported as an NLRP3 inhibitor and is used in studies of inflammation, apoptosis, and neurodegeneration. Supplied as a 10 MG solid, it is intended for laboratory research use only and not for human or clinical applications.
  • Monoterpenoid indole alkaloid with reported NLRP3 inhibitory activity.
  • Useful for studying inflammation, apoptosis, and neurodegenerative processes.
  • Molecular formula C21H24N2O2; molecular weight 336.43 g·mol-1.
  • Supplied as a 10 MG solid for laboratory research.
  • For research use only, not for human or clinical use.