Learn More
Medchemexpress LLC HY-N1431 5mg Medchemexpress, Tabersonine CAS:4429-63-4 Purity:>98%
SDP
Catalog No. 501970337
Encompass_Preferred
Medchemexpress, HY-N1431 5mg Tabersonine CAS:4429-63-4 Tabersonine is an indole alkaloid mainly isolated from Catharanthus roseus. Tabersonine disrupts Aβ(1-42) aggregation and ameliorates Aβ aggregate-induced cytotoxicity. Tabersonine has anti-inflammatory activities and acts as a potential therapeutic candidate for the treatment of ALI/ARDS. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0337 Supplier Medchemexpress LLC Supplier No. HYN14315MG
Medchemexpress, HY-N1431 5mg Tabersonine CAS:4429-63-4 Tabersonine is an indole alkaloid mainly isolated from Catharanthus roseus. Tabersonine disrupts Aβ(1-42) aggregation and ameliorates Aβ aggregate-induced cytotoxicity. Tabersonine has anti-inflammatory activities and acts as a potential therapeutic candidate for the treatment of ALI/ARDS. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.