Learn More
Cayman Chemical Tabersonine, Purity: ≥98%, CAS: 4429-63-4, 10 mg
SDP
Catalog No. 502934894
Encompass_Preferred
An indole alkaloid; inhibits Aβ42 fibril formation and disintegrates preformed Aβ42 fibrils in vitro at 10 μM; an intermediate in the biosynthesis of vindoline
Catalog No. 50-293-4894 Supplier Cayman Chemical Supplier No. 2684810 MG
An indole alkaloid; inhibits Aβ42 fibril formation and disintegrates preformed Aβ42 fibrils in vitro at 10 μM; an intermediate in the biosynthesis of vindoline