Learn More
UNITED SCOPE LLC DBA AMSCOPE AmScope New Microscope Table Stand with Top & Bottom LED Lights
SDP
Catalog No. NC3367605
Encompass_Preferred
A Super Large Size Table Stand with two LED Lights for Stereo Microscopes3" (76mm) Collar12" (305 mm) High Pillar1-1/4" (32mm) Crossing Diameter of Pillar12.5"x10"x0.75" (320x280x20mm) Solid Cast Steel Base PlateWeight: 10 lbs (5.5 kg)
Catalog No. NC3367605 Supplier UNITED SCOPE LLC DBA AMSCOPE Supplier No. TS130LED
A Super Large Size Table Stand with two LED Lights for Stereo Microscopes3" (76mm) Collar12" (305 mm) High Pillar1-1/4" (32mm) Crossing Diameter of Pillar12.5"x10"x0.75" (320x280x20mm) Solid Cast Steel Base PlateWeight: 10 lbs (5.5 kg)