Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Table-Top Tripod Magnifier
SCI_ED

Catalog No. S78988
Change view
Click to view available options
Magnification:
4x
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification
S78988 4x
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

CHOKING HAZARD—Small parts. Not for children under 3 yrs.

Catalog No. S78988 Supplier Learning Resources, Inc Supplier No. LER2905
Add to Cart
Add to Cart

CHOKING HAZARD—Small parts. Not for children under 3 yrs.

Large 4X magnifier is hands-free.

  • Inquisitive kids will get a closer look at insects, rocks, grass—even their own fingers—with this scientific tool
  • Simply rest this 4X magnifier on the table, floor, or ground to bring objects into view
  • Dimensions: 9 dia. x 7 in. (22.5 x 17.5cm)

Specifications

Product Type Magnifier, Table Top Tripod
Height (Metric) 17.78 cm
Diameter (Metric) Outer 22.86 cm
Height (English) 7 in.
For Use With (Application) To get a closer look at insects, rocks, grass, fingers
Diameter (English) Outer 9 in.
Magnification 4x
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.