Learn More
Perkin Elmer US LLC Tablet Measuring System - Fixed Plate
Catalog No. 502113470
Encompass_Preferred
The Measuring System for Tablets holds non-flat tablets, capsules, and pills in compression. It can be used with the fluid bath or the humidity chamber as well as the standard DMA furnace.
Catalog No. 50-211-3470 Supplier Perkin Elmer US LLC Supplier No. N5331244
The Measuring System for Tablets holds non-flat tablets, capsules, and pills in compression. It can be used with the fluid bath or the humidity chamber as well as the standard DMA furnace.