Learn More
Perkin Elmer US LLC Tablet Measuring System - Moving Plate
Catalog No. 502113472
Encompass_Preferred
The Measuring System for Tablets holds non-flat tablets, capsules, and pills in compression. It can be used with the fluid bath or the humidity chamber as well as the standard DMA furnace.
Catalog No. 50-211-3472 Supplier Perkin Elmer US LLC Supplier No. N5331243
The Measuring System for Tablets holds non-flat tablets, capsules, and pills in compression. It can be used with the fluid bath or the humidity chamber as well as the standard DMA furnace.