Learn More
American International Electric TABLETOP IMPULSE SEALER 12 INC
Catalog No. NC2058138
Encompass_Preferred
Tabletop impulse sealer 12 inches, 8 mil Thicknes, 5mm Width and 850W
Catalog No. NC2058138 Supplier American International Electric Supplier No. AIE305HIM
Tabletop impulse sealer 12 inches, 8 mil Thicknes, 5mm Width and 850W