Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tacalcitol (1α,24R-dihydroxyvitamin D3) | 57333-96-7 | MFCD00866623 | 99.7% | 416.64 | C27H44O3 | 10 MG
SDP

Catalog No. 5000296940
Encompass_Preferred

Tacalcitol is a synthetic vitamin D3 analogue supplied as a solid research chemical for studies of bone growth, calcium regulation, inflammation, cancer biology, and dermatological research. The material is a white to off-white powder with high purity and is suitable for in vitro applications when dissolved in appropriate solvents.

  • Synthetic vitamin D3 analogue for research applications.
  • High purity (99.7%).
  • White to off-white solid powder.
  • Available in small pack sizes, including 10 mg vials.
  • Soluble in DMSO at ≥100 mg/mL for in vitro use.
  • Store at 4°C, protect from light, and keep under inert atmosphere for long-term stability.

Catalog No. 50-002-96940 Supplier Medchemexpress LLC Supplier No. HY3233710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tacalcitol is a synthetic vitamin D3 analogue supplied as a solid research chemical for studies of bone growth, calcium regulation, inflammation, cancer biology, and dermatological research. The material is a white to off-white powder with high purity and is suitable for in vitro applications when dissolved in appropriate solvents.

  • Synthetic vitamin D3 analogue for research applications.
  • High purity (99.7%).
  • White to off-white solid powder.
  • Available in small pack sizes, including 10 mg vials.
  • Soluble in DMSO at ≥100 mg/mL for in vitro use.
  • Store at 4°C, protect from light, and keep under inert atmosphere for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.