Learn More
Medchemexpress LLC Tacalcitol monohydrate | 93129-94-3 | 99.9% | 434.65 | 5 MG
SDP
Catalog No. 5000401324
Encompass_Preferred
Tacalcitol monohydrate promotes normal bone growth by regulating calcium ions. It also inhibits cancer cell proliferation and migration. This compound is suitable for research related to inflammation, cancer, and skin diseases.
  • Promotes normal bone growth through calcium ion regulation.
  • Inhibits cancer cell proliferation and migration.
  • Inhibits the synthesis of IL-6 and IL-8 in nasal polyp fibroblast cultures in a dose-dependent manner.
  • Induces cell differentiation in normal human keratinocytes.
  • Inhibits the growth of normal and psoriatic human keratinocytes.
  • Inhibits DNA synthesis in normal human keratinocytes.
  • Strongly reduces the proliferation of human glioblastoma.
  • Inhibits human glioblastoma migration.
  • Significantly inhibits TPA-induced cutaneous inflammation and improves epidermal differentiation in hairless mice.
Catalog No. 50-004-01324 Supplier Medchemexpress LLC Supplier No. HY323385MG
Tacalcitol monohydrate promotes normal bone growth by regulating calcium ions. It also inhibits cancer cell proliferation and migration. This compound is suitable for research related to inflammation, cancer, and skin diseases.
  • Promotes normal bone growth through calcium ion regulation.
  • Inhibits cancer cell proliferation and migration.
  • Inhibits the synthesis of IL-6 and IL-8 in nasal polyp fibroblast cultures in a dose-dependent manner.
  • Induces cell differentiation in normal human keratinocytes.
  • Inhibits the growth of normal and psoriatic human keratinocytes.
  • Inhibits DNA synthesis in normal human keratinocytes.
  • Strongly reduces the proliferation of human glioblastoma.
  • Inhibits human glioblastoma migration.
  • Significantly inhibits TPA-induced cutaneous inflammation and improves epidermal differentiation in hairless mice.