Learn More
Aviva Systems Biology Corp TACC3 Antibody - middle region (ARP52149_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52149_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TACC3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human TACC3. Alternate names: ERIC1, MGC117382, MGC133242, ERIC-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-650