Learn More
Cayman Chemical TaccalonlIde AJ 10mg
SDP
Catalog No. 502942732
Encompass_Preferred
A taccalonolide with anticancer activity; enhances microtubule polymerization in a cell-free assay from 5-30 µM; inhibits the proliferation of HeLa cells (IC50 = 4 nM)
Catalog No. 50-294-2732 Supplier Cayman Chemical Supplier No. 4122310 MG
A taccalonolide with anticancer activity; enhances microtubule polymerization in a cell-free assay from 5-30 µM; inhibits the proliferation of HeLa cells (IC50 = 4 nM)