Learn More
Medchemexpress LLC Taccalonolide A | 108885-68-3 | 5 MG
SDP
Catalog No. 5000420457
Encompass_Preferred
Taccalonolide A is a microtubule stabilizer, a steroid isolated from Tacca chantrieri. It exhibits cytotoxic and antimalarial activities, causing G2-M accumulation, Bcl-2 phosphorylation, and initiating apoptosis. It is effective in vitro against cell lines that overexpress P-glycoprotein (Pgp) and multidrug resistance protein 7 (MRP7), with an IC50 of 622 nM for SK-OV-3 cells.
  • Functions as a microtubule stabilizer
  • Isolated from Tacca chantrieri
  • Exhibits cytotoxic and antimalarial properties
  • Causes G2-M accumulation and Bcl-2 phosphorylation
  • Initiates apoptosis
  • Effective against cell lines overexpressing P-glycoprotein (Pgp)
  • Effective against cell lines overexpressing multidrug resistance protein 7 (MRP7)
Catalog No. 50-004-20457 Supplier Medchemexpress LLC Supplier No. HYN24165MG
Taccalonolide A is a microtubule stabilizer, a steroid isolated from Tacca chantrieri. It exhibits cytotoxic and antimalarial activities, causing G2-M accumulation, Bcl-2 phosphorylation, and initiating apoptosis. It is effective in vitro against cell lines that overexpress P-glycoprotein (Pgp) and multidrug resistance protein 7 (MRP7), with an IC50 of 622 nM for SK-OV-3 cells.
  • Functions as a microtubule stabilizer
  • Isolated from Tacca chantrieri
  • Exhibits cytotoxic and antimalarial properties
  • Causes G2-M accumulation and Bcl-2 phosphorylation
  • Initiates apoptosis
  • Effective against cell lines overexpressing P-glycoprotein (Pgp)
  • Effective against cell lines overexpressing multidrug resistance protein 7 (MRP7)