Learn More
Medchemexpress LLC HY-N4208 5mg Medchemexpress, Taccalonolide AJ CAS:1349904-82-0 Purity:>98%
SDP
Catalog No. 501970435
Encompass_Preferred
Medchemexpress, HY-N4208 5mg Taccalonolide AJ CAS:1349904-82-0 Taccalonolide AJ is a semi-synthesis compound with cellular microtubule stabilizing activity. Taccalonolide AJ exhibits high potency antiproliferative activity against cancer cells, with an IC50 of 4.2 nM for HeLa cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0435 Supplier Medchemexpress LLC Supplier No. HYN42085MG
Medchemexpress, HY-N4208 5mg Taccalonolide AJ CAS:1349904-82-0 Taccalonolide AJ is a semi-synthesis compound with cellular microtubule stabilizing activity. Taccalonolide AJ exhibits high potency antiproliferative activity against cancer cells, with an IC50 of 4.2 nM for HeLa cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.