Learn More
Echelon Biosciences Research Labs TACE Substrate (Fluorogenic) - 5 mg, CAS#: 396716-14-6
SDP
Catalog No. 501908893
Encompass_Preferred
TACE Substrate (Fluorogenic) - 5mg , Dabcyl-Leu-Ala-Gln-Ala-Val-Arg-Ser-Ser-Ser-Arg-EDANS Category/Application: Enzyme Substrates This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-190-8893 Supplier Echelon Biosciences Research Labs Supplier No. 860785MG
TACE Substrate (Fluorogenic) - 5mg , Dabcyl-Leu-Ala-Gln-Ala-Val-Arg-Ser-Ser-Ser-Arg-EDANS Category/Application: Enzyme Substrates This item is non returnable