Learn More
Medchemexpress LLC Tacedinaline (N-acetyldinaline) | 112522-64-2 | MFCD00866266 | 269.30 | C15H15N3O2 | 10 MG
SDP
Catalog No. 501873909
Encompass_Preferred
Tacedinaline (N-acetyldinaline) is a benzamide-class histone deacetylase (HDAC) inhibitor supplied as an analytical standard for research and analytical applications. It inhibits recombinant HDAC1, HDAC2, and HDAC3 with reported IC50 values of 0.9 μM, 0.9 μM, and 1.2 μM, respectively. The compound has molecular formula C15H15N3O2, molecular weight 269.30, and CAS number 112522-64-2.
  • Intended for research and analytical applications.
  • Provides a biochemical assay reference for HDAC activity.
  • Reported activity against HDAC1-3 at micromolar concentrations.
  • Available in small pack sizes suitable for analytical use.
Catalog No. 50-187-3909 Supplier Medchemexpress LLC Supplier No. HY5093410MG
Tacedinaline (N-acetyldinaline) is a benzamide-class histone deacetylase (HDAC) inhibitor supplied as an analytical standard for research and analytical applications. It inhibits recombinant HDAC1, HDAC2, and HDAC3 with reported IC50 values of 0.9 μM, 0.9 μM, and 1.2 μM, respectively. The compound has molecular formula C15H15N3O2, molecular weight 269.30, and CAS number 112522-64-2.
  • Intended for research and analytical applications.
  • Provides a biochemical assay reference for HDAC activity.
  • Reported activity against HDAC1-3 at micromolar concentrations.
  • Available in small pack sizes suitable for analytical use.