Learn More
TARGETMOL CHEMICALS INC Tacedinaline 200MG
SDP
Catalog No. 502878337
Encompass_Preferred
Also available in 1 mL, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Tacedinaline (CI994) is a selective class I HDAC inhibitor with potential antineoplastic activity. Purity 98.37%
Catalog No. 50-287-8337 Supplier TARGETMOL CHEMICALS INC Supplier No. T1888200MG
Also available in 1 mL, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Tacedinaline (CI994) is a selective class I HDAC inhibitor with potential antineoplastic activity. Purity 98.37%