Learn More
Selleck Chemical LLC Tacedinaline10mg
SDP
Catalog No. 502173105
Encompass_Preferred
Tacedinaline (CI994, PD-123654, GOE-5549, Acetyldinaline) is a selective class I HDAC inhibitor with IC50 of 0.9, 0.9, 1.2, and >20 ?M for human HDAC 1, 2, 3, and 8, respectively. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3105 Supplier Selleck Chemical LLC Supplier No. S281810MG
Tacedinaline (CI994, PD-123654, GOE-5549, Acetyldinaline) is a selective class I HDAC inhibitor with IC50 of 0.9, 0.9, 1.2, and >20 ?M for human HDAC 1, 2, 3, and 8, respectively. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.