Learn More
Biocare Medical, LLC Tacha's Bluing Solution, 10X
SDP
Catalog No. 5082883
Encompass_Preferred
Tacha Bluing Solution is highly stable and is designed for bluing hematoxylin stained nuclei. It can be used for both paraffin and frozen sections.
Catalog No. 50-828-83 Supplier Biocare Medical, LLC Supplier No. HTBLUMX
Tacha Bluing Solution is highly stable and is designed for bluing hematoxylin stained nuclei. It can be used for both paraffin and frozen sections.