Learn More
Biocare Medical, LLC Tacha's Bluing Solution
SDP
Catalog No. 5082882
Encompass_Preferred
Tacha Bluing Solution is highly stable and is designed for bluing hematoxylin stainednuclei. It can be used for both paraffin and frozen sections.
Catalog No. 50-828-82 Supplier Biocare Medical, LLC Supplier No. HTBLUM
Tacha Bluing Solution is highly stable and is designed for bluing hematoxylin stainednuclei. It can be used for both paraffin and frozen sections.