Learn More
Cole-Parmer Cole-Parmer Convertible Laser Tachometer with LCD Display
Catalog No. 50993598 Shop All Cole-Parmer Products
Encompass
Select the LCD model for use in bright light or the LED model for easy reading in dim light. A laser spot makes these tachometers easy to aim. Use the memory button to recall last, maximum, minimum, and ten stored readings. Tachometers feature an overrange indicator and low battery indicator.
Catalog No. 50-993-598 Supplier Cole-Parmer Supplier No. 8730400
Select the LCD model for use in bright light or the LED model for easy reading in dim light. A laser spot makes these tachometers easy to aim. Use the memory button to recall last, maximum, minimum, and ten stored readings. Tachometers feature an overrange indicator and low battery indicator.